CAS RN 1211516-26-5
IUPAC Nome:
4-(pentafluoro-λ⁶-sulfanyl)benzene-1-sulfonyl chloride
Sinonimi:
4-(pentafluoro-λ⁶-sulfanyl)benzenesulfonyl chloride
Peso molecolare (g/mol):
302.66
Formula molecolare:
C6H4ClF5O2S2
InChi Key:
LQEOIYDDOVVHIF-UHFFFAOYSA-N
SMILES:
FS(F)(F)(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
di
1
risultati
1
4-(Pentafluorothio)benzenesulfonyl chloride, 97%, Thermo Scientific™
CAS: 1211516-26-5 Formula molecolare: C6H4ClF5O2S2 Molecular Weight (g/mol): 302.658 Numero MDL: MFCD16652450 InChI Key: LQEOIYDDOVVHIF-UHFFFAOYSA-N Sinonimo: 4-pentafluorosulfur benzenesulfonyl chloride,4-pentafluoro-??-sulfanyl benzenesulfonyl chloride,4-pentafluorothio benzenesulfonyl chloride,4-pentafluoro-??-sulfanyl benzene-1-sulfonyl chloride,4-pentafluoro-lambda-sulfanyl benzene-1-sulfonyl chloride PubChem CID: 66523518 IUPAC Name: 4-(pentafluoro-$l^{6}-sulfanyl)benzenesulfonyl chloride SMILES: C1=CC(=CC=C1S(=O)(=O)Cl)S(F)(F)(F)(F)F