CAS RN 14542-13-3
IUPAC Nome:
2-methoxy-1,3-thiazole
Sinonimi:
2-methoxy-1,3-thiazole
Peso molecolare (g/mol):
115.15
Formula molecolare:
C4H5NOS
InChi Key:
MJJRDTKNLLMJDJ-UHFFFAOYSA-N
SMILES:
COC1=NC=CS1
1
–
1
di
1
risultati
1
2-Methoxythiazole, 98%, Thermo Scientific™
CAS: 14542-13-3 Formula molecolare: C4H5NOS Molecular Weight (g/mol): 115.15 Numero MDL: MFCD01631143 InChI Key: MJJRDTKNLLMJDJ-UHFFFAOYSA-N Sinonimo: 2-methoxythiazole,thiazole, 2-methoxy,2-methoxy thiazole,acmc-209cuy,#,ksc174c3j,2-methoxy-1,3-thiazole PubChem CID: 575451 IUPAC Name: 2-methoxy-1,3-thiazole SMILES: COC1=NC=CS1