CAS RN 16214-27-0
IUPAC Nome:
2,3-dihydro-1H-indene-1,2-dione
Sinonimi:
3H-indene-1,2-dione
Peso molecolare (g/mol):
146.15
Formula molecolare:
C9H6O2
InChi Key:
WFFZGYRTVIPBFN-UHFFFAOYSA-N
SMILES:
O=C1CC2=CC=CC=C2C1=O