CAS RN 1722-10-7
IUPAC Nome:
3-chloro-6-methoxypyridazine
Sinonimi:
3-chloro-6-methoxypyridazine
Peso molecolare (g/mol):
144.56
Formula molecolare:
C5H5ClN2O
InChi Key:
XBJLKXOOHLLTPG-UHFFFAOYSA-N
SMILES:
COC1=CC=C(Cl)N=N1
1
–
2
di
2
risultati
1
3-Chloro-6-methoxypyridazine, 97%
CAS: 1722-10-7 Formula molecolare: C5H5ClN2O Molecular Weight (g/mol): 144.56 Numero MDL: MFCD00006467 InChI Key: XBJLKXOOHLLTPG-UHFFFAOYSA-N PubChem CID: 74403 IUPAC Name: 3-chloro-6-methoxypyridazine SMILES: COC1=CC=C(Cl)N=N1
2
3-Chloro-6-methoxypyridazine, Thermo Scientific™
CAS: 1722-10-7 Formula molecolare: C5H5ClN2O Molecular Weight (g/mol): 144.56 Numero MDL: MFCD00006467 InChI Key: XBJLKXOOHLLTPG-UHFFFAOYSA-N PubChem CID: 74403 IUPAC Name: 3-chloro-6-methoxypyridazine SMILES: COC1=CC=C(Cl)N=N1