CAS RN 17475-67-1
IUPAC Nome:
tetrakis(4-hydroxypent-3-en-2-one) hafnium
Sinonimi:
hafnium acetylacetonate
Peso molecolare (g/mol):
578.96
Formula molecolare:
C20H32HfO8
InChi Key:
XFRUSXFSJAEXPU-UHFFFAOYSA-N
SMILES:
[Hf].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O
1
–
1
di
1
risultati
1
Hafnium(IV) 2,4-pentanedionate, 97%
CAS: 17475-67-1 Formula molecolare: C20H28HfO8 Molecular Weight (g/mol): 574.92 Numero MDL: MFCD00044997 InChI Key: GXNMJQUZCICHBX-UHFFFAOYSA-N Sinonimo: hafnium iv acetylacetonate,hafnium-2,4-pentanedionate,hafnium; z-4-hydroxypent-3-en-2-one,3z-4-tris 2z-4-oxopent-2-en-2-yl oxy hafnio oxy pent-3-en-2-one PubChem CID: 11330624 IUPAC Name: hafnium;(Z)-4-hydroxypent-3-en-2-one SMILES: [Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O