CAS RN 1869-24-5
IUPAC Nome:
2-(trifluoromethyl)benzene-1-sulfonamide
Sinonimi:
2-(trifluoromethyl)benzenesulfonamide
Peso molecolare (g/mol):
225.19
Formula molecolare:
C7H6F3NO2S
InChi Key:
AFFPZJFLSDVZBV-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=CC=C1C(F)(F)F
1
–
1
di
1
risultati
1
2-(Trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific™
CAS: 1869-24-5 Formula molecolare: C7H6F3NO2S Molecular Weight (g/mol): 225.185 Numero MDL: MFCD00042420 InChI Key: AFFPZJFLSDVZBV-UHFFFAOYSA-N Sinonimo: 2-trifluoromethyl benzenesulfonamide,2-trifluoromethyl benzene-1-sulfonamide,2-trifluoromethyl benzenesulphonamide,o-trifluoromethylbenzenesulfonamide,benzenesulfonamide, 2-trifluoromethyl,pubchem11781,acmc-20a48s,ksc174g3h,2-trifluoromethylbenzenesul fonamide PubChem CID: 2778018 IUPAC Name: 2-(trifluoromethyl)benzenesulfonamide SMILES: C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)N