CAS RN 19014-29-0
IUPAC Nome:
{4-[4-(dihydroxyboranyl)phenoxy]phenyl}boronic acid
Sinonimi:
4-[4-(dihydroxyboranyl)phenoxy]phenylboronic acid
Peso molecolare (g/mol):
257.84
Formula molecolare:
C12H12B2O5
InChi Key:
DFPCWEXYGPRULG-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(OC2=CC=C(C=C2)B(O)O)C=C1