CAS RN 19777-66-3
IUPAC Nome:
propane-1,2-diamine dihydrochloride
Sinonimi:
propylenediamine dihydrochloride
Peso molecolare (g/mol):
147.04
Formula molecolare:
C3H12Cl2N2
InChi Key:
AEIAMRMQKCPGJR-UHFFFAOYNA-N
SMILES:
Cl.Cl.CC(N)CN