CAS RN 207974-18-3
IUPAC Nome:
1-[2-fluoro-5-(trifluoromethyl)phenyl]propan-1-one
Sinonimi:
1-[2-fluoro-5-(trifluoromethyl)phenyl]propan-1-one
Peso molecolare (g/mol):
220.17
Formula molecolare:
C10H8F4O
InChi Key:
SCKAMDGXXQFMFY-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=C(F)C=CC(=C1)C(F)(F)F
1
–
1
di
1
risultati
1
2'-Fluoro-5'-(trifluoromethyl)propiophenone, 97%, Thermo Scientific™
CAS: 207974-18-3 Formula molecolare: C10H8F4O Molecular Weight (g/mol): 220.17 Numero MDL: MFCD00061260 InChI Key: SCKAMDGXXQFMFY-UHFFFAOYSA-N Sinonimo: 2-fluoro-5-trifluoromethyl propiophenone,2'-fluoro-5'-trifluoromethyl propiophenone,1-2-fluoro-5-trifluoromethyl phenyl propan-1-one,acmc-1cich,5-fluoro-2-trifluoromethyl propiophenone,1-2-fluoranyl-5-trifluoromethyl phenyl propan-1-one,1-2-fluoro-5-trifluoromethyl phenyl-1-propanone # PubChem CID: 605675 IUPAC Name: 1-[2-fluoro-5-(trifluoromethyl)phenyl]propan-1-one SMILES: CCC(=O)C1=C(F)C=CC(=C1)C(F)(F)F