CAS RN 210241-69-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
D-erythro-Imidazoleglycerol Phosphate Monohydrate (~90%), TRC
CAS: 210241-69-3 Formula molecolare: C6 H11 N2 O6 P . H2 O Molecular Weight (g/mol): 256.15 IUPAC Name: [(2R,3S)-2,3-dihydroxy-3-(4H-imidazol-4-yl)propyl] dihydrogen phosphate;hydrate SMILES: O.O[C@H](COP(=O)(O)O)[C@@H](O)C1C=NC=N1