CAS RN 216144-18-2
IUPAC Nome:
2-bromo-1-[4-(pyrrolidin-1-yl)phenyl]ethan-1-one
Sinonimi:
2-bromo-1-[4-(pyrrolidin-1-yl)phenyl]ethanone
Peso molecolare (g/mol):
268.15
Formula molecolare:
C12H14BrNO
InChi Key:
WLRIUCQUMKJOGT-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC=C(C=C1)N1CCCC1
1
–
1
di
1
risultati
1
2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%
CAS: 216144-18-2 Formula molecolare: C12H14BrNO Molecular Weight (g/mol): 268.154 Numero MDL: MFCD01075690 InChI Key: WLRIUCQUMKJOGT-UHFFFAOYSA-N Sinonimo: 2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone,alpha-bromo-4-1-pyrrolidino acetophenone,2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone,2-bromo-1-4-1-pyrrolidinyl phenyl ethanone,ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl,2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one,2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide,2-bromo-1-4-pyrrolidinylphenyl ethan-1-one,acmc-209fm8 PubChem CID: 3681785 IUPAC Name: 2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone SMILES: C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr