CAS RN 23491-52-3
IUPAC Nome:
2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-1,3-benzodiazol-2-yl]-1H-1,3-benzodiazole
Sinonimi:
bisbenzimide
Peso molecolare (g/mol):
452.56
Formula molecolare:
C27H28N6O
InChi Key:
PRDFBSVERLRRMY-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NC2=CC=C(C=C2N1)N1CCN(C)CC1