CAS RN 24083-19-0
IUPAC Nome:
4-(dodecyloxy)benzaldehyde
Sinonimi:
4-(dodecyloxy)benzaldehyde
Peso molecolare (g/mol):
290.45
Formula molecolare:
C19H30O2
InChi Key:
ZBEGLEYBWGNZJA-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCOC1=CC=C(C=O)C=C1
1
–
1
di
1
risultati
1
4-n-Dodecyloxybenzaldehyde, 98%
CAS: 24083-19-0 Formula molecolare: C19H30O2 Molecular Weight (g/mol): 290.447 Numero MDL: MFCD00043523 InChI Key: ZBEGLEYBWGNZJA-UHFFFAOYSA-N Sinonimo: p-dodecyloxybenzaldehyde,4-dodecyloxy benzaldehyde,4-dodecyloxybenzaldehyde,4-n-dodecyloxybenzaldehyde,p-n-dodecoxy benzaldehyde,p-dodecyloxy benzaldehyde,4-dodecyloxy-benzaldehyde,p-lauryloxy benzaldehyde,1-4-formylphenoxy dodecane PubChem CID: 141069 IUPAC Name: 4-dodecoxybenzaldehyde SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)C=O