CAS RN 24131-31-5
IUPAC Nome:
[3,5-bis(benzyloxy)phenyl]methanol
Sinonimi:
[3,5-bis(benzyloxy)phenyl]methanol
Peso molecolare (g/mol):
320.39
Formula molecolare:
C21H20O3
InChi Key:
MHHXKZKHZMSINU-UHFFFAOYSA-N
SMILES:
OCC1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
1
–
3
di
3
risultati
1
3,5-Dibenzyloxybenzyl Alcohol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,5-Dibenzyloxybenzyl Alcohol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,5-Dibenzyloxybenzyl Alcohol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.