CAS RN 282727-18-8
IUPAC Nome:
3-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Sinonimi:
3-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Peso molecolare (g/mol):
341.45
Formula molecolare:
C9H9IO2Zn
InChi Key:
KNTSJWIWLNBYJL-UHFFFAOYSA-M
SMILES:
[Zn+]I.CCOC(=O)C1=C[C-]=CC=C1