CAS RN 29799-07-3
IUPAC Nome:
4-(adamantan-1-yl)phenol
Sinonimi:
4-(adamantan-1-yl)phenol
Peso molecolare (g/mol):
228.34
Formula molecolare:
C16H20O
InChi Key:
KZMYFIUFUAOZHP-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C12CC3CC(CC(C3)C1)C2
1
–
3
di
3
risultati
1
4-(1-Adamantyl)phenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(1-Adamantyl)phenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(1-Adamantyl)phenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.