CAS RN 29840-73-1
IUPAC Nome:
2-bromopyridine-4-carboxamide
Sinonimi:
2-bromopyridine-4-carboxamide
Peso molecolare (g/mol):
201.02
Formula molecolare:
C6H5BrN2O
InChi Key:
KCELTDZFDHPTBI-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=NC(Br)=C1
1
–
1
di
1
risultati
1
2-Bromopyridine-4-carboxamide, 97%
CAS: 29840-73-1 Formula molecolare: C6H5BrN2O Molecular Weight (g/mol): 201.023 Numero MDL: MFCD00234156 InChI Key: KCELTDZFDHPTBI-UHFFFAOYSA-N PubChem CID: 4625407 IUPAC Name: 2-bromopyridine-4-carboxamide SMILES: C1=CN=C(C=C1C(=O)N)Br