CAS RN 331686-36-3
IUPAC Nome:
2-ethylhexanoic acid propan-2-ol iron
Sinonimi:
2-ethylhexanoic acid isopropyl alcohol iron
Peso molecolare (g/mol):
260.16
Formula molecolare:
C11H24FeO3
InChi Key:
VRCGRDMZKVCBSY-UHFFFAOYNA-N
SMILES:
[Fe].CC(C)O.CCCCC(CC)C(O)=O
1
–
1
di
1
risultati
1
Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane, Thermo Scientific™
CAS: 331686-36-3 Formula molecolare: C11H24FeO3 Molecular Weight (g/mol): 260.155 Numero MDL: MFCD00798541 InChI Key: VRCGRDMZKVCBSY-UHFFFAOYSA-N Sinonimo: iron ii 2-ethylhexano-isopropoxide,in hexane,iron ii 2-ethylhexano-isopropoxide w/v in hexane,iron ii 2-ethylhexanoate monoisopropoxide w/v in hexane PubChem CID: 121233721 IUPAC Name: 2-ethylhexanoic acid;iron;propan-2-ol SMILES: CCCCC(CC)C(=O)O.CC(C)O.[Fe]