CAS RN 3468-18-6
IUPAC Nome:
1-(1H-indol-4-yl)methanamine
Sinonimi:
1-(1H-indol-4-yl)methanamine
Peso molecolare (g/mol):
146.19
Formula molecolare:
C9H10N2
InChi Key:
FFBWKPKOXRMLNP-UHFFFAOYSA-N
SMILES:
NCC1=C2C=CNC2=CC=C1
1
–
1
di
1
risultati
1
4-(Aminomethyl)indole, 97%
CAS: 3468-18-6 Formula molecolare: C9H10N2 Molecular Weight (g/mol): 146.19 Numero MDL: MFCD04973297 InChI Key: FFBWKPKOXRMLNP-UHFFFAOYSA-N Sinonimo: 4-aminomethylindole,1h-indol-4-yl methanamine,1h-indole-4-methanamine,4-aminomethyl indole,1h-indol-4-ylmethyl amine,1-1h-indol-4-yl methanamine,c-1h-indol-4-yl-methylamine,indole-4-methylamine,4-aminomethyl-indole,4-aminomethyl-1h-indole PubChem CID: 280302 IUPAC Name: 1H-indol-4-ylmethanamine SMILES: NCC1=C2C=CNC2=CC=C1