CAS RN 3637-01-2
IUPAC Nome:
1-(3,4-dimethylphenyl)ethan-1-one
Sinonimi:
1-(3,4-dimethylphenyl)ethanone
Peso molecolare (g/mol):
148.21
Formula molecolare:
C10H12O
InChi Key:
WPRAXAOJIODQJR-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(C)C(C)=C1
1
–
1
di
1
risultati
1
3',4'-Dimethylacetophenone, 98%
CAS: 3637-01-2 Formula molecolare: C10H12O Molecular Weight (g/mol): 148.21 Numero MDL: MFCD00008743 InChI Key: WPRAXAOJIODQJR-UHFFFAOYSA-N