CAS RN 36821-26-8
IUPAC Nome:
ethyl 2-oxopyrrolidine-3-carboxylate
Sinonimi:
ethyl 2-oxopyrrolidine-3-carboxylate
Peso molecolare (g/mol):
157.17
Formula molecolare:
C7H11NO3
InChi Key:
FHHMWJMUMOYGQW-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1CCNC1=O
1
–
1
di
1
risultati
1
Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals
CAS: 36821-26-8 Formula molecolare: C7H11NO3 Molecular Weight (g/mol): 157.169 Numero MDL: MFCD06796534 InChI Key: FHHMWJMUMOYGQW-UHFFFAOYSA-N PubChem CID: 12699000 IUPAC Name: ethyl 2-oxopyrrolidine-3-carboxylate SMILES: CCOC(=O)C1CCNC1=O