CAS RN 453-14-5
IUPAC Nome:
1,3-difluoropropan-2-one
Sinonimi:
1,3-difluoropropan-2-one
Peso molecolare (g/mol):
94.06
Formula molecolare:
C3H4F2O
InChi Key:
HKIPCXRNASWFRU-UHFFFAOYSA-N
SMILES:
FCC(=O)CF
1
–
1
di
1
risultati
1
1,3-Difluoroacetone, 98%
CAS: 453-14-5 Formula molecolare: C3H4F2O Molecular Weight (g/mol): 94.061 Numero MDL: MFCD07368665 InChI Key: HKIPCXRNASWFRU-UHFFFAOYSA-N Sinonimo: 1,3-difluoroacetone,2-propanone, 1,3-difluoro,1,3-difluoro-2-propanone,ch2f 2co,1,3-bis fluoranyl propan-2-one,fluoromethyl ketone,sym-difluoroacetone,sgqdlibrbrwsp@,1, 3-difluoroacetone,1,3-difluoro acetone PubChem CID: 228397 IUPAC Name: 1,3-difluoropropan-2-one SMILES: C(C(=O)CF)F