CAS RN 493038-93-0
IUPAC Nome:
2-(3-chloro-5-fluorophenyl)acetonitrile
Sinonimi:
2-(3-chloro-5-fluorophenyl)acetonitrile
Peso molecolare (g/mol):
169.58
Formula molecolare:
C8H5ClFN
InChi Key:
GWPLYSLPTCBNDL-UHFFFAOYSA-N
SMILES:
FC1=CC(Cl)=CC(CC#N)=C1
1
–
1
di
1
risultati
1
3-Chloro-5-fluorophenylacetonitrile, 97%, Thermo Scientific™
CAS: 493038-93-0 Formula molecolare: C8H5ClFN Molecular Weight (g/mol): 169.583 Numero MDL: MFCD04038259 InChI Key: GWPLYSLPTCBNDL-UHFFFAOYSA-N Sinonimo: 3-chloro-5-fluorophenylacetonitrile,2-3-chloro-5-fluorophenyl acetonitrile,3-chloro-5-fluorobenzyl cyanide,3-chloro-5-fluorophenyl acetonitrile,2-3-chloro-5-fluoro-phenyl acetonitrile,pubchem4280,ksc494a3d,3-chloro-5-fluorobenzylcyanide,2-5-chloro-3-fluorophenyl ethanenitrile PubChem CID: 2734836 IUPAC Name: 2-(3-chloro-5-fluorophenyl)acetonitrile SMILES: C1=C(C=C(C=C1F)Cl)CC#N