CAS RN 50530-12-6
IUPAC Nome:
10-bromodecanoic acid
Sinonimi:
10-bromodecanoic acid
Peso molecolare (g/mol):
251.16
Formula molecolare:
C10H19BrO2
InChi Key:
PGVRSPIEZYGOAD-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCBr
1
–
1
di
1
risultati
1
10-Bromodecanoic acid, 95%
CAS: 50530-12-6 Formula molecolare: C10H19BrO2 Molecular Weight (g/mol): 251.164 Numero MDL: MFCD00014388 InChI Key: PGVRSPIEZYGOAD-UHFFFAOYSA-N Sinonimo: 10-bromo-decanoic acid,decanoic acid, 10-bromo,10-bromanyldecanoic acid,10-bromodecanoicacid,acmc-1aonn,10-bromodecanoic acid.,.omega.-bromodecanoic acid,ghl.pd_mitscher_leg0.481,ksc489q1r,10-bromodecanoic acid PubChem CID: 142712 IUPAC Name: 10-bromodecanoic acid SMILES: C(CCCCC(=O)O)CCCCBr