CAS RN 53324-38-2
IUPAC Nome:
4-bromo-3-nitroaniline
Sinonimi:
4-bromo-3-nitroaniline
Peso molecolare (g/mol):
217.02
Formula molecolare:
C6H5BrN2O2
InChi Key:
PITHQPMZWKZGRZ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(Br)C(=C1)[N+]([O-])=O