CAS RN 584-20-3
CAS RN 584-20-3
IUPAC Nome:
(3S)-3-azaniumyl-4,4,4-trifluorobutanoate
Sinonimi:
(3S)-3-ammonio-4,4,4-trifluorobutanoate
Peso molecolare (g/mol):
157.09
Formula molecolare:
C4H6F3NO2
InChi Key:
XRXQOEWWPPJVII-REOHCLBHSA-N
SMILES:
[NH3+][C@@H](CC([O-])=O)C(F)(F)F
1
–
1
di
1
risultati
1
3-Amino-4,4,4-trifluorobutyric acid, 97%
CAS: 584-20-3 Formula molecolare: C4H6F3NO2 Molecular Weight (g/mol): 157.09 Numero MDL: MFCD00041413 InChI Key: XRXQOEWWPPJVII-REOHCLBHSA-N PubChem CID: 342970 IUPAC Name: 3-amino-4,4,4-trifluorobutanoic acid SMILES: [NH3+][C@@H](CC([O-])=O)C(F)(F)F