CAS RN 612-94-2
IUPAC Nome:
2-phenylnaphthalene
Sinonimi:
naphthalene, 2-phenyl-
Peso molecolare (g/mol):
204.27
Formula molecolare:
C16H12
InChi Key:
TURIHPLQSRVWHU-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1