CAS RN 64742-65-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
CAS: 64742-65-0 Numero MDL: MFCD00147539