CAS RN 6786-36-3
IUPAC Nome:
1-phenyloctadecan-1-one
Sinonimi:
1-phenyloctadecan-1-one
Peso molecolare (g/mol):
344.58
Formula molecolare:
C24H40O
InChi Key:
DBLXXVQTWJFJFI-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
1
–
1
di
1
risultati
1
n-Octadecanophenone, 99%, Thermo Scientific™
CAS: 6786-36-3 Formula molecolare: C24H40O Molecular Weight (g/mol): 344.583 Numero MDL: MFCD00010700 InChI Key: DBLXXVQTWJFJFI-UHFFFAOYSA-N Sinonimo: octadecanophenone,stearophenone,n-octadecanophenone,heptadecyl phenyl ketone,1-octadecanone, 1-phenyl,1-phenyl-1-octadecanone # PubChem CID: 81244 IUPAC Name: 1-phenyloctadecan-1-one SMILES: CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1