CAS RN 72955-82-9
IUPAC Nome:
[2-(2-methoxyphenoxy)ethyl](methyl)amine
Sinonimi:
[2-(2-methoxyphenoxy)ethyl](methyl)amine
Peso molecolare (g/mol):
181.24
Formula molecolare:
C10H15NO2
InChi Key:
VOIPEKOXTFLQNY-UHFFFAOYSA-N
SMILES:
CNCCOC1=CC=CC=C1OC
1
–
1
di
1
risultati
1
2-(2-Methoxyphenoxy)-N-methylethylamine, 96%, Thermo Scientific™
CAS: 72955-82-9 Formula molecolare: C10H15NO2 Molecular Weight (g/mol): 181.235 Numero MDL: MFCD07400597 InChI Key: VOIPEKOXTFLQNY-UHFFFAOYSA-N Sinonimo: 2-2-methoxyphenoxy-n-methylethanamine,2-2-methoxyphenoxy-n-methylethylamine,2-2-methoxyphenoxy ethyl methyl amine,2-2-methoxy-phenoxy-ethyl-methyl-amine,2-2-methoxyphenoxy ethyl methylamine,2-2-methoxyphenoxy-n-methyl-ethanamine,2-2-methoxyphenoxy ;-n-methylethanamine,ethanamine,2-2-methoxyphenoxy-n-methyl PubChem CID: 6485441 IUPAC Name: 2-(2-methoxyphenoxy)-N-methylethanamine SMILES: CNCCOC1=CC=CC=C1OC