CAS RN 749927-69-3
IUPAC Nome:
4-bromo-2-fluoro-N-methylbenzamide
Sinonimi:
4-bromo-2-fluoro-N-methylbenzamide
Peso molecolare (g/mol):
232.05
Formula molecolare:
C8H7BrFNO
InChi Key:
BAJCFNRLEJHPTQ-UHFFFAOYSA-N
SMILES:
CNC(=O)C1=C(F)C=C(Br)C=C1
1
–
2
di
2
risultati
1
4-Bromo-2-fluoro-N-methylbenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Bromo-2-fluoro-N-methylbenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.