CAS RN 776-76-1
IUPAC Nome:
methyldiphenylsilyl
Sinonimi:
methyldiphenylsilyl
Peso molecolare (g/mol):
197.33
Formula molecolare:
C13H13Si
InChi Key:
OKHRRIGNGQFVEE-UHFFFAOYSA-N
SMILES:
C[Si](C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
di
1
risultati
1
Methyldiphenylsilane, 97%
CAS: 776-76-1 Formula molecolare: C13H13Si Molecular Weight (g/mol): 197.332 Numero MDL: MFCD00008255 InChI Key: OKHRRIGNGQFVEE-UHFFFAOYSA-N Sinonimo: methyldiphenylsilane,diphenylmethylsilane,silane, methyldiphenyl,benzene, 1,1'-methylsilylene bis,methyldiphenylsilyl,methyl diphenyl silicon,methyl-di phenyl silicon,silane, methyldiphenyl-, PubChem CID: 6327660 IUPAC Name: methyl(diphenyl)silicon SMILES: C[Si](C1=CC=CC=C1)C2=CC=CC=C2