CAS RN 777-37-7
IUPAC Nome:
1-chloro-4-nitro-2-(trifluoromethyl)benzene
Sinonimi:
1-chloro-4-nitro-2-(trifluoromethyl)benzene
Peso molecolare (g/mol):
225.55
Formula molecolare:
C7H3ClF3NO2
InChi Key:
HQROXDLWVGFPDE-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F
1
–
1
di
1
risultati
1
2-Chloro-5-nitrobenzotrifluoride, 98%
CAS: 777-37-7 Formula molecolare: C7H3ClF3NO2 Molecular Weight (g/mol): 225.55 Numero MDL: MFCD00007296 InChI Key: HQROXDLWVGFPDE-UHFFFAOYSA-N Sinonimo: 2-chloro-5-nitrobenzotrifluoride,1-chloro-4-nitro-2-trifluoromethyl benzene,4-chloro-3-trifluoromethyl nitrobenzene,benzene, 1-chloro-4-nitro-2-trifluoromethyl,3-trifluoromethyl-4-chloronitrobenzene,2-trifluoromethyl-4-nitrochlorobenzene,2-chloro-5-nitro-a,a,a-trifluorotoluene,4-nitro-2-trifluoromethyl chlorobenzene,1-chloro-2-trifluoromethyl-4-nitrobenzene,5-nitro-2-chlorobenzotrifluoride PubChem CID: 61213 IUPAC Name: 1-chloro-4-nitro-2-(trifluoromethyl)benzene SMILES: [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F