CAS RN 869807-57-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Andropanolide, MedChemExpress
MedChemExpress Andrographolide (Andro) is a small antagonist for NF-κB activation by covalent modifying reduced cysteine 62 of p50.