CAS RN 876014-27-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
(1S,5R,6R)-5-Hydroxy-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1S,5R,6R)-5-Hydroxy-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.