CAS RN 87680-47-5
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
N-Desiminoethyl,-N-(4-nitrobenzyloxycarbonyl) Panipenem 4-Nitrobenzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Desiminoethyl,-N-(4-nitrobenzyloxycarbonyl) Panipenem 4-Nitrobenzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Desiminoethyl,-N-(4-nitrobenzyloxycarbonyl) Panipenem 4-Nitrobenzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.