CAS RN 88-24-4
IUPAC Nome:
2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Sinonimi:
2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Peso molecolare (g/mol):
368.56
Formula molecolare:
C25H36O2
InChi Key:
GPNYZBKIGXGYNU-UHFFFAOYSA-N
SMILES:
CCC1=CC(=C(O)C(CC2=C(O)C(=CC(CC)=C2)C(C)(C)C)=C1)C(C)(C)C
1
–
3
di
3
risultati
1
6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.