CAS RN 880468-89-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
(R)-tert-Butyl 1-(Benzylamino)-3-methoxy-1-oxopropan-2-ylcarbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-tert-Butyl 1-(Benzylamino)-3-methoxy-1-oxopropan-2-ylcarbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-tert-Butyl 1-(Benzylamino)-3-methoxy-1-oxopropan-2-ylcarbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.