CAS RN 88058-69-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
[1R-[1Alpha(R*),2Beta]]-Alpha-Methoxy-benzeneacetic Acid 1,2,3,4-Tetrahydro-7-methoxy-2-[(1-oxopropyl)amino]-1-naphthalenyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[1R-[1Alpha(R*),2Beta]]-Alpha-Methoxy-benzeneacetic Acid 1,2,3,4-Tetrahydro-7-methoxy-2-[(1-oxopropyl)amino]-1-naphthalenyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.