CAS RN 88165-26-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Methyl (3aR,7R,7aS)-3a,6,7,7a-Tetrahydro-7-hydroxy-2,2-dimethyl-1,3-benzodioxole-5-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl (3aR,7R,7aS)-3a,6,7,7a-Tetrahydro-7-hydroxy-2,2-dimethyl-1,3-benzodioxole-5-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl (3aR,7R,7aS)-3a,6,7,7a-Tetrahydro-7-hydroxy-2,2-dimethyl-1,3-benzodioxole-5-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.