CAS RN 88404-22-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
N-(tert-Butyloxy)carbonyl-L-thyroxine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(tert-Butyloxy)carbonyl-L-thyroxine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(tert-Butyloxy)carbonyl-L-thyroxine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.