CAS RN 885702-34-1
IUPAC Nome:
4-bromo-2-chloropyrimidine
Sinonimi:
4-bromo-2-chloropyrimidine
Peso molecolare (g/mol):
193.43
Formula molecolare:
C4H2BrClN2
InChi Key:
FBEBVAQOMVWORE-UHFFFAOYSA-N
SMILES:
ClC1=NC=CC(Br)=N1
1
–
1
di
1
risultati
1
4-Bromo-2-chloropyrimidine, 98%, Thermo Scientific Chemicals
CAS: 885702-34-1 Formula molecolare: C4H2BrClN2 Molecular Weight (g/mol): 193.43 Numero MDL: MFCD08272196 InChI Key: FBEBVAQOMVWORE-UHFFFAOYSA-N Sinonimo: pyrimidine, 4-bromo-2-chloro-9ci,pyrimidine, 4-bromo-2-chloro,2-chloro-4-bromopyrimidine PubChem CID: 45117394 IUPAC Name: 4-bromo-2-chloropyrimidine SMILES: ClC1=NC=CC(Br)=N1