CAS RN 886499-64-5
IUPAC Nome:
1-(bromomethyl)-4,5-difluoro-2-methoxybenzene
Sinonimi:
1-(bromomethyl)-4,5-difluoro-2-methoxybenzene
Peso molecolare (g/mol):
237.04
Formula molecolare:
C8H7BrF2O
InChi Key:
PRLKVWCFDNQAED-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(F)C=C1CBr
1
–
1
di
1
risultati
1
4,5-Difluoro-2-methoxybenzyl bromide, 97%, Thermo Scientific™
CAS: 886499-64-5 Formula molecolare: C8H7BrF2O Molecular Weight (g/mol): 237.044 Numero MDL: MFCD06660220 InChI Key: PRLKVWCFDNQAED-UHFFFAOYSA-N Sinonimo: 4,5-difluoro-2-methoxybenzyl bromide,1-bromomethyl-4,5-difluoro-2-methoxybenzene PubChem CID: 17750791 IUPAC Name: 1-(bromomethyl)-4,5-difluoro-2-methoxybenzene SMILES: COC1=CC(=C(C=C1CBr)F)F