CAS RN 90101-20-5
IUPAC Nome:
tert-butyl N-(4-methylpyridin-2-yl)carbamate
Sinonimi:
tert-butyl N-(4-methylpyridin-2-yl)carbamate
Peso molecolare (g/mol):
208.26
Formula molecolare:
C11H16N2O2
InChi Key:
SABMAMDNSRZJBK-UHFFFAOYSA-N
SMILES:
CC1=CC(NC(=O)OC(C)(C)C)=NC=C1
1
–
1
di
1
risultati
1
2-(Boc-amino)-4-methylpyridine, 97%, Thermo Scientific™
CAS: 90101-20-5 Formula molecolare: C11H16N2O2 Molecular Weight (g/mol): 208.261 Numero MDL: MFCD07776933 InChI Key: SABMAMDNSRZJBK-UHFFFAOYSA-N Sinonimo: 4-methyl-pyridin-2-yl-carbamic acid tert-butyl ester,tert-butyl 4-methylpyridin-2-yl carbamate,tert-butyl n-4-methylpyridin-2-yl carbamate,2-boc-amino-4-methylpyridine,tert-butyl 4-methylpyridin-2-ylcarbamate,4-methylpyridin-2-yl carbamic acid tert-butyl ester,carbamic acid, 4-methyl-2-pyridinyl-, 1,1-dimethylethyl ester,ambkkkkk140,tert-butyl 4-methyl-2-pyridinylcarbamate PubChem CID: 11063648 IUPAC Name: tert-butyl N-(4-methylpyridin-2-yl)carbamate SMILES: CC1=CC(=NC=C1)NC(=O)OC(C)(C)C