CAS RN 94109-09-8
CAS RN 94109-09-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Tris(tridecyl) Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tris(tridecyl) Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tris(tridecyl) Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.