CAS RN 942206-16-8
IUPAC Nome:
2',6-dibromo-3,4'-bipyridine
Sinonimi:
2',6-dibromo-3,4'-bipyridine
Peso molecolare (g/mol):
313.98
Formula molecolare:
C10H6Br2N2
InChi Key:
LGVJILYMUFZGLC-UHFFFAOYSA-N
SMILES:
BrC1=NC=C(C=C1)C1=CC(Br)=NC=C1
1
–
1
di
1
risultati
1
2',6-Dibromo-3,4'-bipyridine, 95%, Thermo Scientific™
CAS: 942206-16-8 Formula molecolare: C10H6Br2N2 Molecular Weight (g/mol): 313.98 Numero MDL: MFCD09037424 InChI Key: LGVJILYMUFZGLC-UHFFFAOYSA-N Sinonimo: 2',6-dibromo-3,4'-bipyridine,6,2'-dibromo-3,4'-bipyridine,2-bromo-5-2-bromopyridin-4-yl pyridine,6,2'-dibromo-3,4' bipyridine PubChem CID: 42553011 IUPAC Name: 2-bromo-4-(6-bromopyridin-3-yl)pyridine SMILES: BrC1=NC=C(C=C1)C1=CC(Br)=NC=C1