CAS RN 944070-25-1
CAS RN 944070-25-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-(methyl)-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-(methyl)-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.