CAS RN 944538-49-2
CAS RN 944538-49-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
(R)-Bethanechol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-Bethanechol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.