CAS RN 945725-50-8
CAS RN 945725-50-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Tocris Bioscience™ TASP 0277308
Potent and selective S1P1 receptor antagonist; inhibits angiogenesis