CAS RN 947-02-4
CAS RN 947-02-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Phosfolan, TRC
CAS: 947-02-4 Formula molecolare: C7 H14 N O3 P S2 Molecular Weight (g/mol): 255.29 Sinonimo: Imidocarbonic acid, phosphonodithio-, cyclic ethylene P,P-diethyl ester (7CI,8CI),Phosphoramidic acid, 1,3-dithiolan-2-ylidene-, diethyl ester (9CI),(Diethoxyphosphinyl)dithioimidocarbonic acid cyclic ethylene ester,2-[Diethoxyphosphinylimino]-1,3-dithiolane,AC 47031,American Cyanamide 47031,CL 47031,Cyolane,Diethyl 1,3-dithiolan-2-ylidenephosphoramidate,EI 47031,ENT 25,830,Phosfolan,Phospholan IUPAC Name: N-diethoxyphosphoryl-1,3-dithiolan-2-imine SMILES: CCOP(=O)(OCC)N=C1SCCS1